Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFKM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102782 Supplier Invitrogen™ Supplier No. PA5102782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PFKM.

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PFKM
Gene Accession No. P08237, P47857, P47858
Gene Alias 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; AI131669; ATP-dependent 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; GSD7; HAMAP-Rule:MF_03184}; PFK; PFK1; PFK-1; Pfk4; Pfk-4; Pfka; PFK-A; PFKM; Pfk-m; PFKX; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-A; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); phosphohexokinase; phosphohexokinase {ECO:0000255; PPP1R122; protein phosphatase 1, regulatory subunit 122
Gene Symbols PFKM
Host Species Rabbit
Immunogen A synthesized peptide derived from human PFKM(Accession P08237), corresponding to amino acid residues F49-L99.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18642, 5213, 65152
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.