Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFKP Antibody (OTI1D6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273344MFV450
Description
PFKP Monoclonal antibody specifically detects PFKP in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
PFKP | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI1D6 | |
Flow Cytometry | |
6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase | |
Full length human recombinant protein of human PFKP(NP_002618) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
5214 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction