Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFKP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584295 Supplier Invitrogen™ Supplier No. PA584295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MECVQMTQDV QKAMDERRFQ DAVRLRGRSF AGNLNTYKRL AIKLPDDQIP KTNCNVAVIN V Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The AGC kinase group consists of 63 kinases including the cyclic nucleotide-regulated protein kinase (PKA & PKG) family, the diacylglycerol-activated/phospholipid-dependent protein kinase C (PKC) family, the related to PKA and PKC (RAC/Akt) protein kinase family, the kinases that phosphorylate G protein-coupled receptors family (ARK), and the kinases that phosphorylate ribosomal protein S6 family (RSK).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKP
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PFKP
Gene Accession No. Q01813
Gene Alias 1200015H23Rik; 6-phosphofructo-1-kinase, C-type; 6-phosphofructokinase type C; 6-phosphofructokinase, platelet type; 9330125N24Rik; ATP:D-fructose 6-phosphate 1-phosphotransferase; ATP-dependent 6-phosphofructokinase, platelet type; ATP-dependent 6-phosphofructokinase, platelet type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; EC 2.7.1.11; HAMAP-Rule:MF_03184}; K6PP; PFK 1; PFK1; Pfkc; PFK-C; PFKF; PFKP; PFK-P; Phosphofructo-1-kinase isozyme C; Phosphofructokinase; phosphofructokinase 1; phosphofructokinase, platelet; Phosphofructokinase, platelet type; phosphohexokinase; phosphohexokinase {ECO:0000255
Gene Symbols PFKP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PFKP. Recombinant protein control fragment (Product #RP-105043).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5214
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.