Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFN2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2H7C12, Proteintech
SDP

Catalog No. 501736239 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6239 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6239 Supplier Proteintech Group Inc Supplier No. 600942IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MW of this protein is 15 kDa, and this antibody specifically recognizes the 15 kDa protein.

Profilin 2 is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals.

Specifications

Antigen PFN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H7C12
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PFN2
Gene Accession No. P35080, Q9EPC6, Q9JJV2
Gene Alias D3S1319E, PFL, PFN2, Prifilin 2, PROF2, profilin 2, Profilin II
Gene Symbols Pfn2
Host Species Mouse
Immunogen PFN2 Fusion Protein Ag5043
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100621523, 18645, 5217, 81531
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.