Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ PFTE Stopper with Extraction Nut



Stopper has a nylon extraction nut permitting easy removal of the stopper from the glass outer joint.
$36.60 - $67.80
Specifications
Color | White |
---|---|
Material | PTFE |
For Use With (Application) | Organic Synthesis |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
31-501-905
![]() ![]() |
Synthware™
S531018 |
10/18 |
Each for $36.60
|
|
|||||
31-501-906
![]() ![]() |
Synthware™
S531420 |
14/20 |
Each for $43.20
|
|
|||||
31-501-907
![]() ![]() |
Synthware™
S531922 |
19/22 |
Each for $42.00
|
|
|||||
31-501-908
![]() ![]() |
Synthware™
S532440 |
24/40 |
Each for $67.80
|
|
|||||
Specifications
White | |
PTFE | |
Organic Synthesis | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title