Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFTK1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5106962

Catalog No. PIPA5106962


Only null left
Add to Cart

Description

Description

Sequence homology: Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

CDK14 (PFTK1)/cyclin Y is a member of the CDC2-related protein kinase family which is expressed primarily in the postnatal and adult nervous system. CDK14 is expressed at high levels in brain, pancreas, kidney, heart, testis, and ovary. CDK14 amino acid residue ser119 is required for its interaction with all four 14-3-3 isoforms, while ser120 may contribute slightly to CDK14 binding. CDK14 acts as a cyclin-dependent kinase that regulates cell cycle progression and cell proliferation. Overexpression of CDK14 predicts resistance to chemotherapy in patients with esophageal squamous cell carcinoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PFTK1
Polyclonal
Unconjugated
Cdk14
Cdk14; Cell division protein kinase 14; cyclin dependent kinase 14; cyclin-dependent kinase 14; hPFTAIRE1; Kiaa0834; mKIAA0834; PFTAIRE protein kinase 1; PFTAIRE1; Pftk1; Serine/threonine-protein kinase PFTAIRE-1
Rabbit
Affinity Chromatography
RUO
18647
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
O35495
Cdk14
Synthetic peptide located within the following region: SDLPPRLWEL TDMSSIFTVP NVRLQPEAGE SMRAFGKNNS YGKSLSNSKH.
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.