Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFTK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PFTK1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3513
![]() ![]() |
Proteintech Group Inc
216121AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to PFTK1.
PFTK1, a member of the CDC2/CDKX subfamily of Ser/Thr protein kinases, may play a role in meiosis as well as in neuron differentiation and/or function It is highly expressed in brain, pancreas, kidney, heart, testis and ovary, and also detected at lower levels in other tissues except in spleen and thymus where expression is minimal.Specifications
PFTK1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O35495, O94921 | |
18647, 362316, 5218 | |
PFTK1 Fusion Protein Ag16259 | |
Primary | |
-20°C | |
CDK14 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDK14, hPFTAIRE1, KIAA0834, PFTAIRE protein kinase 1, PFTAIRE1, PFTK1 | |
Cdk14 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title