Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFTK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553383 Supplier Invitrogen™ Supplier No. PA553383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRSLHVVWNR LGRVPEAEDL ASQMLKGFPR DRVSAQEALV HDYFSALPSQ LYQLPDEESL FTVSGVRLKP EMCDLLASYQ KGHHPA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFTK2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDK15
Gene Accession No. Q96Q40
Gene Alias ALS2CR7; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 7; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 7 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 7 protein homolog; Cdk15; Cell division protein kinase 15; cyclin dependent kinase 15; cyclin-dependent kinase 15; frizzled family receptor 7; Fzd7; PFTAIRE protein kinase 2; PFTAIRE2; PFTK2; serine/threonine protein kinase; serine/threonine-protein kinase ALS2CR7; Serine/threonine-protein kinase PFTAIRE-2
Gene Symbols CDK15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PFTK2. Recombinant protein control fragment (Product #RP-91449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.