Learn More
Invitrogen™ PGA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114247
Description
Immunogen sequence: ISDTNQIFGL SETEPGSFLY YAPFDGILGL AYPSISSSGA TPVFDNIWNQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 83%; Pig: 100%; Rabbit: 100%; Rat: 83%.
PGA3 (Pepsinogen A3) is a Protein Coding gene. Diseases associated with PGA3 include Atrophic Gastritis and Gastritis. Among its related pathways are Collagen chain trimerization and Metabolism of proteins. Gene Ontology (GO) annotations related to this gene include aspartic-type endopeptidase activity. An important paralog of this gene is PGA4.
Specifications
| PGA3 | |
| Polyclonal | |
| Unconjugated | |
| PGA3 | |
| pepsin A-3; Pepsinogen 3; pepsinogen 3, group I (pepsinogen A); pepsinogen A3; pepsinogen-3; PGA3; PGA4; PGA5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 643834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P0DJD8 | |
| PGA3 | |
| Synthetic peptide directed towards the middle region of Human PGA3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.