Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120544 Supplier Invitrogen™ Supplier No. PA5120544
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: C6, U-251MG, U-87MG, SGC-7901, Mouse brain, Rat brain. The target is usually found in the following locations: Secreted. Immunogen sequence: VDEQPLENYL DMEYFGTIGI GTPAQDFTVV FDTGSSNLWV PSVYCSSLAC TNHNRFNPED SSTYQSTSET VSITYGTGSM TGILGYDTVQ VGGISDTNQI FGLSETEPGS FLYYAPFD.

PGA3 (Pepsinogen A3) is a Protein Coding gene. Diseases associated with PGA3 include Atrophic Gastritis and Gastritis. Among its related pathways are Collagen chain trimerization and Metabolism of proteins. Gene Ontology (GO) annotations related to this gene include aspartic-type endopeptidase activity. An important paralog of this gene is PGA4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGA3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PGA3
Gene Accession No. P0DJD8
Gene Alias pepsin A-3; Pepsinogen 3; pepsinogen 3, group I (pepsinogen A); pepsinogen A3; pepsinogen-3; PGA3; PGA4; PGA5
Gene Symbols PGA3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 63-180 of human PGA3 (NP_001073275.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643834
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.