Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAM1/2/4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100774
Description
PGAM1/2/4 Polyclonal specifically detects PGAM1/2/4 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
PGAM1/2/4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
PGAM1, PGAMA, PGAM-B, phosphoglycerate mutase 1 | |
Goat | |
29 kDa | |
0.1 mg | |
Stem Cell Markers | |
5223 | |
Human, Mouse, Rat, Pig | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 0.05-0.15 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:28000 | |
NP_002620 | |
PGAM1 | |
Peptide with sequence C-KAMEAVAAQGKAKK, from the C Terminus of the protein sequence according to NP_002620.1; NP_000281.2; NP_001025062.1. | |
Affinity Purified | |
RUO | |
Primary | |
Please note this antibody is expected to recognize the products of 3 highly similar genes. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction