Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAM1 Antibody (1G4), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259418AF700
This item is not returnable.
View return policy
Description
PGAM1 Monoclonal antibody specifically detects PGAM1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PGAM1 | |
Monoclonal | |
Alexa Fluor 700 | |
BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B | |
Recombinant human PGAM1 (1-254aa) purified from E. coli | |
0.1 mL | |
Stem Cell Markers | |
5223 | |
Store at 4°C in the dark. | |
IgG2a λ |
Western Blot, ELISA, Immunofluorescence | |
1G4 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction