Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAM1 Monoclonal Antibody (GT256)
GREENER_CHOICE

Catalog No. PIMA527770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527770 Supplier Invitrogen™ Supplier No. MA527770
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain Predicted Reactivity: Xenopus laevis (84%), Chicken (92%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombinant human PGAM protein, fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT256
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PGAM1
Gene Accession No. P18669, P25113, Q9DBJ1
Gene Alias 2310050F24Rik; BPG-dependent PGAM 1; CDABP0006; epididymis secretory protein Li 35; HEL-S-35; PGAM1; Pgam-1; PGAMA; PGAM-B; Pgmut; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase isozyme B; RP11-452K12.8; similar to phosphoglycerate mutase
Gene Symbols PGAM1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human PGAM1. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18648, 24642, 5223
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.