Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAM2 Antibody (5A7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259410F
This item is not returnable.
View return policy
Description
PGAM2 Monoclonal antibody specifically detects PGAM2 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
PGAM2 | |
Monoclonal | |
FITC | |
BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
Recombinant human PGAM2 (1-253aa) purified from E. coli | |
0.1 mL | |
Cancer, Proteases & Other Enzymes, Signal Transduction | |
5224 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA, Immunofluorescence | |
5A7 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction