Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PGAM2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PGAM2 Polyclonal specifically detects PGAM2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PGAM2 | |
Polyclonal | |
Rabbit | |
Cancer, Proteases & Other Enzymes, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
PGAM2 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P15259 | |
5224 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PMDEKHPYYNSISKERRYAGLK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title