Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAM2 Antibody (OTI4E9), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273358APCCY7
Description
PGAM2 Monoclonal specifically detects PGAM2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
PGAM2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
PGAM2 | |
Full length recombinant protein of human PGAM2 (NP_000028) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2a |
Flow Cytometry | |
OTI4E9 | |
Flow Cytometry | |
BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
Mouse | |
Affinity Purified | |
Cancer, Proteases & Other Enzymes, Signal Transduction | |
5224.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction