Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM2 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
SDP

Catalog No. 502540930
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0930 30 μL
50-167-4309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0930 Supplier OriGene Supplier No. TA503408S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGAM2 Monoclonal antibody specifically detects PGAM2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PGAM2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ANAPC2
Gene Accession No. Q8BZQ7, Q9UJX6
Gene Alias BPG-dependent PGAM 2; D14Mgh1; EGK_13739; GSD10; Muscle-specific phosphoglycerate mutase; Pgam2; PGAMM; PGAM-M; Pgmut; Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human ANAPC2 produced in HEk293 cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29882, 99152
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.