Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584444 Supplier Invitrogen™ Supplier No. PA584444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMDEKHPYYN SISKERRYAG LK Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.

Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). PGAM2 encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pgam2
Gene Accession No. P15259
Gene Alias BPG-dependent PGAM 2; D14Mgh1; EGK_13739; GSD10; Muscle-specific phosphoglycerate mutase; Pgam2; PGAMM; PGAM-M; Pgmut; Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit
Gene Symbols Pgam2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PGAM2 (Product #RP-103369)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.