Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PGAM4 Monoclonal Antibody (OTI1H7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527361
Description
PGAM4 Monoclonal Antibody for Western Blot, IHC (P)
Phosphoglyceric acid mutase is widely distributed in mammalian tissues where it catalyzes the reversible reaction of 3-phosphoglycerate to 2-phosphoglycerate in the glycolytic pathway.
Specifications
PGAM4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N0Y7 | |
PGAM4 | |
Full length human recombinant protein of PGAM4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H7 | |
Unconjugated | |
PGAM4 | |
dJ1000K24.1; MGC88743; OTTHUMP00000059414; PGAM1; PGAM2; PGAM3; PGAM4; PGAMA; PGAM-B; PGAMM; phosphoglycerate mutase 4; phosphoglycerate mutase family 3; phosphoglycerate mutase family member 4; phosphoglycerate mutase processed protein; probable phosphoglycerate mutase 4; RP11-452K12.8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
441531 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction