Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM4 Mouse anti-Human, Clone: OTI1A1, lyophilized, TrueMAB™
SDP

Catalog No. 501670309
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0309 Supplier OriGene Supplier No. CF810791
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Phosphoglyceric acid mutase is widely distributed in mammalian tissues where it catalyzes the reversible reaction of 3-phosphoglycerate to 2-phosphoglycerate in the glycolytic pathway.

Specifications

Antigen PGAM4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PGAM4
Gene Accession No. Q8N0Y7
Gene Alias PGAM-B, PGAM1, PGAM3, dJ1000K24.1
Gene Symbols PGAM4
Host Species Mouse
Immunogen Full length human recombit protein of human PGAM4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441531
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.