Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571806
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571806 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571806 Supplier Invitrogen™ Supplier No. PA571806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGAM4 Polyclonal Antibody for Western Blot, IHC (P)

Phosphoglyceric acid mutase is widely distributed in mammalian tissues where it catalyzes the reversible reaction of 3-phosphoglycerate to 2-phosphoglycerate in the glycolytic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PGAM4
Gene Accession No. Q8N0Y7
Gene Alias dJ1000K24.1; MGC88743; OTTHUMP00000059414; PGAM1; PGAM2; PGAM3; PGAM4; PGAMA; PGAM-B; PGAMM; phosphoglycerate mutase 4; phosphoglycerate mutase family 3; phosphoglycerate mutase family member 4; phosphoglycerate mutase processed protein; probable phosphoglycerate mutase 4; RP11-452K12.8
Gene Symbols PGAM4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 25-53 amino acids from the N-terminal region of human PGAM4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.