Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAM5 Monoclonal Antibody (CL0624)
GREENER_CHOICE

Catalog No. PIMA524616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524616 Supplier Invitrogen™ Supplier No. MA524616
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: NWDRREPLSL INVRKRNVES GEEELASKLD HYKAKATRHI FLIRHSQYHV DGSLEKDRTL TPLGREQAEL TGLRLASLGL KFNKIV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%. Binds to an epitope located within the peptide sequence NWDRREPLSLINVRK as determined by overlapping synthetic peptides.

Displays phosphatase activity for serine/threonine residues, and, dephosphorylates and activates MAP3K5 kinase. Has apparently no phosphoglycerate mutase activity. May be regulator of mitochondrial dynamics. Substrate for a KEAP1-dependent ubiquitin ligase complex. Contributes to the repression of NFE2L2-dependent gene expression. Acts as a central mediator for programmed necrosis induced by TNF, by reactive oxygen species and by calcium ionophore. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL0624
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGAM5
Gene Accession No. Q96HS1
Gene Alias 2610528A17Rik; Bcl-XL-binding protein v68; BXLBV68; PGAM family member 5, mitochondrial serine/threonine protein phosphatase; PGAM family member 5, serine/threonine protein phosphatase, mitochondrial; PGAM5; Phosphoglycerate mutase family member 5; RGD1312028; serine/threonine-protein phosphatase PGAM5, mitochondrial
Gene Symbols PGAM5
Host Species Mouse
Immunogen Recombinant protein fragment of Human PGAM5 (Product #RP-95712).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.