Learn More
Invitrogen™ PGAM5 Recombinant Rabbit Monoclonal Antibody (7J3A5)

Description
Immunogen sequence: GARPGPGVWD PNWDRREPLS LINVRKRNVE SGEEELASKL DHYKAKATRH IFLIRHSQYH VDGSLEKDRT LTPLGREQAE LTGLRLASLG LKFNKIV.
Specifications
Specifications
| Antigen | PGAM5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 7J3A5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | PGAM5 |
| Gene Accession No. | Q96HS1 |
| Gene Alias | 2610528A17Rik; Bcl-XL-binding protein v68; BXLBV68; PGAM family member 5, mitochondrial serine/threonine protein phosphatase; PGAM family member 5, serine/threonine protein phosphatase, mitochondrial; PGAM5; Phosphoglycerate mutase family member 5; RGD1312028; serine/threonine-protein phosphatase PGAM5, mitochondrial |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.