Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564091 Supplier Invitrogen™ Supplier No. PA564091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FHFEYTVATD CGVPNYLPSV SSAIG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 80%.

PGAP2 is involved in the lipid remodeling steps of GPI-anchor maturation. Required for stable expression of GPI-anchored proteins at the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGAP2
Gene Accession No. Q9UHJ9
Gene Alias cell wall biogenesis 43 N-terminal homolog; CWH43-N; FGF receptor activating protein 1; FGF receptor-activating protein 1; FRAG1; HPMRS3; mental retardation, non-syndromic, autosomal recessive, 21; MRT17; MRT21; PGAP2; post-GPI attachment to proteins 2; post-GPI attachment to proteins factor 2
Gene Symbols PGAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGAP2. Recombinant protein control fragment (Product #RP-100066).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.