Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGAP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18669225UL
Description
PGAP3 Polyclonal specifically detects PGAP3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PGAP3 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
AGLA546, CAB2COS16 homolog, Gene coamplified with ERBB2 protein, MGC9753hCOS16, PER1, per1-like domain containing 1, PERLD1PER1-like domain-containing protein 1, post-GPI attachment to proteins 3, post-GPI attachment to proteins factor 3, PP1498 | |
Rabbit | |
Affinity Purified | |
RUO | |
93210 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PGAP3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SQGDREPVYRDCVLQCEEQNCSGGALNHFRSRQPIYMSLAGWTCRDDCKYECMWVTVGLYLQEGHKVPQFHGKWP | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction