Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PGAP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553451
Description
Immunogen sequence: SQGDREPVYR DCVLQCEEQN CSGGALNHFR SRQPIYMSLA GWTCRDDCKY ECMWVTVGLY LQEGHKVPQF HGKWP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchors proteins. Required for phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI (Probable).
Specifications
PGAP3 | |
Polyclonal | |
Unconjugated | |
PGAP3 | |
AGLA546; CAB2; COS16 homolog; gene coamplified with ERBB2 protein; hCOS16; per1-like domain containing 1; PER1-like domain-containing protein 1; PERLD1; PGAP3; post-GPI attachment to proteins 3; post-GPI attachment to proteins factor 3; PP1498; UNQ546/PRO1100 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
93210 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FM1 | |
PGAP3 | |
Recombinant protein corresponding to Human PGAP3. Recombinant protein control fragment (Product #RP-91223). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction