Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208480
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8480 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8480 Supplier Fabgennix Inc Supplier No. PGAP3101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGAP3 Polyclonal antibody specifically detects PGAP3 in Human, Mouse, Rat samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen PGAP3
Applications Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PGAP3
Gene Accession No. A2A559, Q96FM1
Gene Alias AGLA546; CAB2; COS16 homolog; gene coamplified with ERBB2 protein; hCOS16; per1-like domain containing 1; PER1-like domain-containing protein 1; PERLD1; PGAP3; post-GPI attachment to proteins 3; post-GPI attachment to proteins factor 3; PP1498; UNQ546/PRO1100
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on human PGAP3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320655, 688174, 93210
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.