Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGBD1 Monoclonal Antibody (2A3)
GREENER_CHOICE

Catalog No. PIMA522767
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522767 Supplier Invitrogen™ Supplier No. MA522767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: NLCGNSAQET VMSLSPMTEE IVTKDRLFKA KQETSEEMEQ SGEASGKPNR ECAPQIPCST PIATERTVAH LNTLKDRHPG DLWARMHISS LEYAAGDITR KGRKKDKARV.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGBD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PGBD1
Gene Accession No. Q96JS3
Gene Alias 4921509E05Rik; Cerebral protein 4; cerebral protein-4; dJ874C20.4; hucep-4; PGBD1; piggyBac transposable element derived 1; piggyBac transposable element-derived protein 1; SCAND4
Gene Symbols PGBD1
Host Species Mouse
Immunogen PGBD1 (NP_115896.1, 241 a.a. ~ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84547
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.