Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552410 Supplier Invitrogen™ Supplier No. PA552410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCVKTYPLES GEEAVTVLEN LETGSGDTGQ QASVYIQGQD MHPMVAEYQG VSLECQSLQL LPGITTLKCE PPQRPQGNPQ EVSGPVPHGS AHLQEKNPRD KAVVPVFNPV RSQTLVKTEE ETAQAVAAEK WSHLSLTRRN LCGNS Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 29%.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGBD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGBD1
Gene Accession No. Q96JS3
Gene Alias 4921509E05Rik; Cerebral protein 4; cerebral protein-4; dJ874C20.4; hucep-4; PGBD1; piggyBac transposable element derived 1; piggyBac transposable element-derived protein 1; SCAND4
Gene Symbols PGBD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGBD1. Recombinant protein control fragment (Product #RP-89353).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.