Learn More
PGBD1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303202AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known.Specifications
PGBD1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96JS3 | |
PGBD1 | |
Between 275 and 325 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PGBD1 | |
HUCEP-4, SCAND4, dJ874C20.4, cerebral protein 4, cerebral protein-4, piggyBac transposable element-derived protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84547 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.