Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGBD2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093427
Description
PGBD2 Polyclonal specifically detects PGBD2 in Human samples. It is validated for Peptide ELISA.Specifications
PGBD2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
piggyBac transposable element derived 2, piggyBac transposable element-derived protein 2 | |
Goat | |
Affinity Purified | |
RUO | |
267002 | |
Human | |
IgG |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:16000 | |
NP_001017434.1 | |
PGBD2 | |
Peptide with sequence C-ESNADTTSQGRRSRR corresponding to internal region according to NP_001017434.1, NP_733843.1 . | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001017434.1; NP_733843.1) | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction