Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGBD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559343 Supplier Invitrogen™ Supplier No. PA559343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLTLVNDLLG QGYCVFLDNF NISPMLFREL HQNRTDAVGT ARLNRKQIPN DLKKRIAKGT TVARFCGELM ALKWCDGKEV TMLSTFHNDT VIEVNNRNGK KTKR Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 19%.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGBD4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGBD4
Gene Accession No. Q96DM1
Gene Alias PGBD4; piggyBac transposable element derived 4; piggyBac transposable element-derived protein 4
Gene Symbols PGBD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGBD4. Recombinant protein control fragment (Product #RP-98175).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161779
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.