Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGBD5 Monoclonal Antibody (7-F8-5)
GREENER_CHOICE

Catalog No. PIMA532886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532886 Supplier Invitrogen™ Supplier No. MA532886
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGBD5 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGBD5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7-F8-5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene PGBD5
Gene Accession No. Q8N414
Gene Alias 2900019M05Rik; AI854313; DKFZp761A0620; FLJ11413; PGBD5; piggyBac domain related protein 5; PiggyBac domain-related protein 5; piggyBac transposable element derived 5; piggyBac transposable element-derived protein 5; PiggyBac transposase 5
Gene Symbols PGBD5
Host Species Mouse
Immunogen Synthetic peptide with Human PGBD5 aa 61-110.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209966, 79605
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.