Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGC-1 beta Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128722AF594
Description
PGC-1 beta Polyclonal antibody specifically detects PGC-1 beta in Human samples. It is validated for Western BlotSpecifications
PGC-1 beta | |
Polyclonal | |
Western Blot | |
DKFZp686C1790, ERRL1, PERCFLJ14284, peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PGC1, PGC1Bgamma, coactivator 1, beta, PPARGC1 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human peroxisome proliferator-activated receptor gamma coactivator 1 beta using the numbering given in entry NP_573570.2 (GeneID 133522). | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, Diabetes Research, Hypoxia, Lipid and Metabolism, Mitochondrial Markers, Neuroscience, Signal Transduction | |
133522 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction