Learn More
PGC-1beta Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302274A

Description
The recommended shelf life for this product is 1 year from date of receipt.
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.Specifications
PGC-1beta | |
Polyclonal | |
Unconjugated | |
PPARGC1B | |
Peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PERC, PGC-1(beta), PGC1B, ERRL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133522 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86YN6 | |
PPARGC1B | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.