Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (JF09-71)
GREENER_CHOICE

Catalog No. PIMA532563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532563 Supplier Invitrogen™ Supplier No. MA532563
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGC1 alpha/beta
Applications Western Blot
Classification Recombinant Monoclonal
Clone JF09-71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Ppargc1a
Gene Accession No. O70343, Q811R2, Q86YN6, Q8VHJ7, Q9QYK2, Q9UBK2
Gene Alias 4631412G21Rik; A830037N07Rik; ENSMUSG00000079510; ERR ligand 1; Errl1; Gm11133; LEM6; Ligand effect modulator 6; ligand effect modulator-6; L-PGC-1alpha; PERC; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; peroxisome proliferator-activated receptor gamma coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1-alpha; Peroxisome proliferator-activated receptor gamma coactivator 1-beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma coactivator-1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; Pgc1; PGC-1; pgc-1 alpha; pgc-1 beta; PGC-1(alpha); PGC-1(beta); PGC1A; PGC-1alpha; PGC-1-alpha; Pgc-1alphaa; PGC1B; PGC1beta; PGC-1beta; PGC-1-beta; PGC-1beta/ERRL1; PGC-1-related estrogen receptor alpha coactivator; PGC-1v; Pgco1; PPAR gamma coactivator variant form; PPAR Gamma Coactivator-1; PPAR gamma coactivator-1beta protein; PPARG coactivator 1 alpha; PPARG coactivator 1 beta; PPARgamma coactivator 1 beta; PPARgamma coactivator 1alpha; PPAR-gamma coactivator 1-alpha; PPAR-gamma coactivator 1-beta; Ppargc1; Ppargc1a; PPARGC-1-alpha; Ppargc1b; PPARGC-1-beta
Gene Symbols Ppargc1a, Ppargc1b
Host Species Rabbit
Immunogen Synthetic peptide within human pgc1 alpha aa 755-798 and aa 269-323.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10891, 133522, 170826, 19017, 291567, 83516
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.