Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGCB Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208495
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8495 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8495 Supplier Fabgennix Inc Supplier No. PGCB201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGCB Polyclonal antibody specifically detects PGCB in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PGCB
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Npr2
Gene Accession No. P16067, P20594, Q6VVW5
Gene Alias AMDM; ANPb; ANP-B; ANPRB; ANPR-B; atrial natriuretic peptide B-type receptor; atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; atrial natriuretic peptide receptor type B; cn; ECDM; GC-B; GC-B2; GC-B3; guanylate cyclase 2B; guanylate cyclase B; guanylyl cyclase-B; GUC2B; GUCY2B; mNPR-B; natriuretic peptide receptor 2; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR2; NPRB; NPR-B; NPRBi; pwe; SNSK
Host Species Rabbit
Immunogen Synthetic C-terminal peptide corresponding to unique amino acid sequences on Particulate Guanylyl Cyclase B protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116564, 230103, 4882
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.