Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548235
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548235 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548235 Supplier Invitrogen™ Supplier No. PA548235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGCD Polyclonal Antibody for Western Blot, IHC (P), Flow

Probably plays a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGCD
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GUCY2D
Gene Accession No. P51839, Q02846
Gene Alias cone rod dystrophy 6; CORD5; CORD6; CYGD; guanylate cyclase 2d; guanylate cyclase 2D, membrane (retina-specific); guanylate cyclase 2D, retinal; Guanylate cyclase, olfactory; guanylyl cyclase 2d; guanylyl cyclase D; GUC1A4; GUC2D; GUCY2D; LCA; LCA1; olfactory guanylyl cyclase GC-D; RCD2; RETGC; RETGC1; RETGC-1; retinal guanylate cyclase 1; retinal guanylyl cyclase 1; Rod outer segment membrane guanylate cyclase; ROSGC; ROS-GC; ROS-GC1
Gene Symbols GUCY2D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 540-570 amino acids from the Central region of human GUCY2D.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113911, 3000
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.