Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGCD Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208501
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8501 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8501 Supplier Fabgennix Inc Supplier No. PGCD401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGCD Polyclonal antibody specifically detects PGCD in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PGCD
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene GUCY2D
Gene Accession No. P51839, Q02846
Gene Alias cone rod dystrophy 6; CORD5; CORD6; CYGD; guanylate cyclase 2d; guanylate cyclase 2D, membrane (retina-specific); guanylate cyclase 2D, retinal; Guanylate cyclase, olfactory; guanylyl cyclase 2d; guanylyl cyclase D; GUC1A4; GUC2D; GUCY2D; LCA; LCA1; olfactory guanylyl cyclase GC-D; RCD2; RETGC; RETGC1; RETGC-1; retinal guanylate cyclase 1; retinal guanylyl cyclase 1; Rod outer segment membrane guanylate cyclase; ROSGC; ROS-GC; ROS-GC1
Host Species Rabbit
Immunogen Synthetic C-terminal peptide corresponding to unique amino acid sequences on Particulate Guanylyl Cyclase D protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113911, 14918, 3000
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.