Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGCF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589697 Supplier Invitrogen™ Supplier No. PA589697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSALIGGETQ MHLLECAHDL KMTDGTYVFV PYDALLYSLP YKHTPYRVLR NNPKLREAYD AVLTITVESQ EKTFYQAFTE AAARGEIPEK LEFDQVSPLF GTIYNSIYFI AQAMNNAMKE NGQAGAASLV QHSRNMQFHG FNQLMRTDSN GNGISEYVIL DTNLKEWELH STYTVDMEME LLRFGGTPIH FPGGRPPRAD AKCWFAEGKI C; Positive Samples: Mouse eye; Cellular Location: Membrane, Single-pass type I membrane protein.

GUCY2F (Guanylate cyclase 2F, retinal) is also named as GUC2F, RETGC2 and belongs to the adenylyl cyclase class-4/guanylyl cyclase family. GUCY2F may play a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction. By in situ hybridization, mRNA encoding GUCY2F is found only in the outer nuclear layer and inner segments of photoreceptor cells. By using synthetic peptide antiserum specific for each RetGC subtype, RetGC-2(GUCY2F) can be distinguished from RetGC-1 as a slightly smaller protein in immunoblots of bovine rod outer segments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGCF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GUCY2F
Gene Accession No. P51842, Q5SDA5
Gene Alias A930002I06; CYGF; GC2; GC-F; guanylate cyclase 2D-like, membrane (retina-specific); guanylate cyclase 2f; Guanylate cyclase 2F, retinal; guanylate cyclase F; GUC2DL; GUC2F; Gucy2f; RETGC2; RETGC-2; Retinal guanylyl cyclase 2; rod outer segment membrane guanylate cyclase 2; ROS-GC2
Gene Symbols GUCY2F
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-460 of human GUCY2F (NP_0015132).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116556, 245650
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.