Learn More
PGCF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA131197
Description
PA1-31197 detects PGCF from rat, mouse, human samples. PA1-31197 has been successfully used in Western blot applications. The PA1-31197 immunogen is: Synthetic peptide for particulate guanylyl cyclase F. All peptides were amidated for conjugation. The PGCF selective antibodies were prepared against a unique peptide selective for only PGCF that maps at or close to the C termi

Specifications
PGCF | |
Polyclonal | |
GUCY2F | |
GUCY2F | |
Synthetic peptide for particulate guanylyl cyclase F. All peptides were amidated for conjugation. The PGCF selective antibodies were prepared against a unique peptide selective for only PGCF that maps at or close to the C terminal region of the protein. ( | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
P51841, Q5SDA5, P51842 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116556, 245650, 2986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.