Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547602
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547602 Supplier Invitrogen™ Supplier No. PA547602
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

PGCP is a metallopeptidase that belongs to the peptidase M28 family. The encoded protein may catalyze the cleavage of dipeptides with unsubstituted terminals into amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGCP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Cpq
Gene Accession No. Q9WVJ3
Gene Alias 1190003P12Rik; 2610034C17Rik; aminopeptidase; blood plasma glutamate carboxypeptidase; carboxypeptidase Q; Cpq; Hematopoietic lineage switch 2; hematopoietic lineage switch 2 related protein; Hls2; hls2-rp; LAL; Lal-1; LCH1; LDP; liver annexin like-1; liver annexin-like protein 1; Lysosomal dipeptidase; Pgcp; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase
Gene Symbols Cpq
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PGCP Lys19-Ser470.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54381
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.