Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGD Antibody (46B3), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259449JF646
This item is not returnable.
View return policy
Description
PGD Monoclonal antibody specifically detects PGD in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PGD | |
Monoclonal | |
Janelia Fluor 646 | |
6PGD, EC 1.1.1.44,6-phosphogluconate dehydrogenase, decarboxylating, PGDH, phosphogluconate dehydrogenase | |
Recombinant human PGD (1-483aa) purified from E. coli | |
0.1 mL | |
Endocrinology, Signal Transduction | |
5226 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunofluorescence | |
46B3 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction