Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD Monoclonal Antibody (2A10G4), Proteintech
SDP

Catalog No. 502333250 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3250 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3250 Supplier Proteintech Group Inc Supplier No. 679161IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGD Monoclonal antibody specifically detects PGD in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PGD
Applications Western Blot
Classification Monoclonal
Clone 2A10G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P52209
Gene Alias 6PGD; PGD; PGDH
Host Species Mouse
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5226
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.