Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGD Recombinant Rabbit Monoclonal Antibody (9A2M7)
GREENER_CHOICE

Catalog No. PIMA542605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542605 Supplier Invitrogen™ Supplier No. MA542605
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, HepG2, Jurkat, Mouse lung, Mouse liver, Mouse spleen, Rat lung, Rat liver, Rat kidney. The target is usually found in the following locations: Cytoplasm.

6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGD
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9A2M7
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene PGD
Gene Accession No. P52209, P85968, Q9DCD0
Gene Alias 0610042A05Rik; 6PGD; 6-phosphogluconate dehydrogenase, decarboxylating; 6-phosphogluconate dehydrogenase, decarboxylating-like; AU019875; C78335; LOC100360180; Pgd; PGDH; phosphogluconate dehydrogenase
Gene Symbols PGD
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360180, 110208, 5226
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.