Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGD2 Synthase/PTGDS Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059911AF594
Description
PGD2 Synthase/PTGDS Polyclonal specifically detects PGD2 Synthase/PTGDS in Human, Mouse, Rat, Bovine, Canine, Feline, Primate, Rabbit samples. It is validated for Western Blot.Specifications
PGD2 Synthase/PTGDS | |
Polyclonal | |
Western Blot | |
Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase | |
Rabbit | |
Affinity Purified | |
Mast Cell Markers, Neuroscience, Signal Transduction | |
5730.0 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PTGDS | |
The epitope recognized by this antibody maps to region within residue 120-190 of human Prostaglandin D Synthase/Lipocalin. [Swiss-ProtNo. P41222] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Feline, Primate, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction