Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGE2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP289043
Description
PGE2 Polyclonal specifically detects PGE2 in Non-species specific samples. It is validated for ELISA.Specifications
PGE2 | |
Polyclonal | |
PBS containing 0.01M sodium azide | |
Rabbit | |
100 μL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
ELISA | |
Unconjugated | |
PG E2, PGE 2, PGE2, ProstaglandinE2 | |
Prostaglandin E2 (PGE2) conjugated to bovine thyroglobulin. | |
RUO | |
Non-species specific |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction