Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pGEM™ Vectors

Catalog No. PRP2161 Shop All Promega Corporation Products
Change view
Click to view available options
Vector:
pGEM-3Z vector
pGEM-3Zf(+) vector
pGEM-4Z vector
pGEM-5Zf(+) vector
pGEM-7Zf(+) vector
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PR-P2161 pGEM-4Z vector
PR-P2271 pGEM-3Zf(+) vector
PR-P2241 pGEM-5Zf(+) vector
PRP2251 pGEM-7Zf(+) vector
PR-P2151 pGEM-3Z vector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PR-P2161 Supplier Promega Supplier No. P2161
Only null left
Add to Cart
Add to Cart

Includes

Vector, 20μg; Bacterial Strain JM109, 500μL

Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters

  • Standard Cloning Vector That can also Efficiently Synthesize RNA In vitro
  • Blue/white screening to easily identify recombinant clones
  • Versatile vector used for standard cloning and in vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Multiple cloning site provides a convenient selection of restriction sites for cloning

Specifications

Promoter SP6, T7
Restriction Site Acc I, Ava I, BamH I, EcoRI, Hinc II, Hind III, Kpn I, Pst I, Sac I, Sal I, Sma I, Sph I, Xba I, Xma I
Content And Storage -30°C to -10°C
Quantity 20 μg
Vector pGEM-4Z vector
For Use With (Application) Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.