Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Pgf (Mouse) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
Pgf (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse PLGF-2 in cell culture supernates, serum and plasma (heparin, EDTA).
Specifications
Specifications
Description | placental growth factor |
Cross Reactivity | Mouse |
Target Species | Mouse |
Gene ID (Entrez) | 18654 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for six months. For long term storage store at -20°C. Avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction