Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGF Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA579813
Description
Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
Specifications
PGF | |
Polyclonal | |
PBS with 5MG BSA and 0.02% sodium azide, 0.02% thimerosal | |
P49763 | |
PGF | |
A synthetic peptide corresponding to a sequence at the C-terminus of human PLGF. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PGF | |
Placenta growth factor, PlGF, PGF, PGFL, PLGF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5228 | |
-20°C | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction